COM448635196 Combi-Blocks, Inc.

3,5-Difluoropyridine 1-oxide

  • MDL number MFCD16036156
  • Cas Number : 210169-07-6
  • Molecular Weight : 131.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PY-1359

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?PY-1359
Availability details for
Quantity
  • Availability
  • Add to Cart