COM448635197 Combi-Blocks, Inc.

3-Chloro-5-fluoropyridine 1-oxide

  • MDL number MFCD16036157
  • Cas Number : 1221793-68-5
  • Molecular Weight : 147.5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PY-1360

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?PY-1360
Availability details for
Quantity
  • Availability
  • Add to Cart