COM448635220 Combi-Blocks, Inc.

3-(Benzyloxy)-6-methylpyridazine

  • MDL number MFCD16036159
  • Cas Number : 6958-54-9
  • Molecular Weight : 200.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PY-1395

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?PY-1395
Availability details for
Quantity
  • Availability
  • Add to Cart