COM448635224 Combi-Blocks, Inc.

Methyl 6-acetamido-3-chloropicolinate

  • MDL number MFCD16660314
  • Cas Number : 1242336-62-4
  • Molecular Weight : 228.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PY-1400

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?PY-1400
Availability details for
Quantity
  • Availability
  • Add to Cart