COM448635230 Combi-Blocks, Inc.

4-Bromo-2-tert-butoxypyridine

  • MDL number MFCD11223214
  • Cas Number : 1086381-36-3
  • Molecular Weight : 230.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PY-1409

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?PY-1409
Availability details for
Quantity
  • Availability
  • Add to Cart