COM448635239 Combi-Blocks, Inc.

3-Bromo-2-(methylthio)-5-(trifluoromethyl)pyridine

  • MDL number MFCD16295130
  • Cas Number : 1226808-64-5
  • Molecular Weight : 272.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PY-1420

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?PY-1420
Availability details for
Quantity
  • Availability
  • Add to Cart