COM448635243 Combi-Blocks, Inc.

3-Bromo-2-(methylthio)pyridine

  • MDL number MFCD12964310
  • Cas Number : 51933-77-8
  • Molecular Weight : 204.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PY-1424

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?PY-1424
Availability details for
Quantity
  • Availability
  • Add to Cart