COM448635285 Combi-Blocks, Inc.

5-Bromo-2-(N-methylsufonylamino)pyrimidine

  • MDL number MFCD16660319
  • Cas Number : 1242336-55-5
  • Molecular Weight : 252.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PY-1481

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?PY-1481
Availability details for
Quantity
  • Availability
  • Add to Cart