COM448635286 Combi-Blocks, Inc.

3-Cyano-2-pyrrolidinopyridine

  • MDL number MFCD07792921
  • Cas Number : 59025-38-6
  • Molecular Weight : 173.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PY-1483

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?PY-1483
Availability details for
Quantity
  • Availability
  • Add to Cart