COM448635294 Combi-Blocks, Inc.

2-Chloro-4-pyrrolidinopyridine

  • MDL number MFCD09032267
  • Cas Number : 874758-84-6
  • Molecular Weight : 182.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PY-1493

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?PY-1493
Availability details for
Quantity
  • Availability
  • Add to Cart