COM448635296 Combi-Blocks, Inc.

2-Chloro-4-morpholinopyridine

  • MDL number MFCD12923071
  • Cas Number : 937202-67-0
  • Molecular Weight : 198.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PY-1495

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?PY-1495
Availability details for
Quantity
  • Availability
  • Add to Cart