COM448635298 Combi-Blocks, Inc.

5-Bromo-2-(N-cyclopentylamino)pyrimidine

  • MDL number MFCD17015844
  • Cas Number : 1207293-60-4
  • Molecular Weight : 242.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PY-1497

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?PY-1497
Availability details for
Quantity
  • Availability
  • Add to Cart