COM448635300 Combi-Blocks, Inc.

5-Bromo-2-(2,2,2-trifluoroethyl)aminopyrimidine

  • MDL number MFCD17015846
  • Cas Number : 1245563-08-9
  • Molecular Weight : 256.0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PY-1499

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?PY-1499
Availability details for
Quantity
  • Availability
  • Add to Cart