COM448635306 Combi-Blocks, Inc.

3-Bromo-2-methyl-6-propoxypyridine

  • MDL number MFCD18783193
  • Cas Number : 1280786-78-8
  • Molecular Weight : 230.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PY-1514

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?PY-1514
Availability details for
Quantity
  • Availability
  • Add to Cart