COM448635315 Combi-Blocks, Inc.

3,5-Dibromo-6-chloropyridin-2-ol

  • MDL number MFCD17015847
  • Cas Number : 6515-39-5
  • Molecular Weight : 287.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PY-1524

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?PY-1524
Availability details for
Quantity
  • Availability
  • Add to Cart