COM448635317 Combi-Blocks, Inc.

5-Bromo-2-cyclopropylaminopyridine

  • MDL number MFCD08706291
  • Cas Number : 885266-96-6
  • Molecular Weight : 213.08

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 5-bromo-N-cyclopropyl-2-pyridinamine
Inchi Code 1S/C8H9BrN2/c9-6-1-4-8(10-5-6)11-7-2-3-7/h1,4-5,7H,2-3H2,(H,10,11)
Inchi Key ITKZPCOJFUVIQO-UHFFFAOYSA-N

Description

Purity 98%
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PY-1533

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?PY-1533
Availability details for
Quantity
  • Availability
  • Add to Cart