COM448635318 Combi-Blocks, Inc.

5-Bromo-2-propylaminopyridine

  • MDL number MFCD09743643
  • Cas Number : 100379-02-0
  • Molecular Weight : 215.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PY-1534

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?PY-1534
Availability details for
Quantity
  • Availability
  • Add to Cart