COM448635319 Combi-Blocks, Inc.

2-(4-Nitrophenylamino)pyridine

  • MDL number MFCD18087748
  • Cas Number : 24068-29-9
  • Molecular Weight : 215.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PY-1535

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?PY-1535
Availability details for
Quantity
  • Availability
  • Add to Cart