COM448635330 Combi-Blocks, Inc.

6-Bromo-5-methyltetrazolo[1,5-a]pyridine

  • MDL number MFCD18087749
  • Cas Number : 1257665-09-0
  • Molecular Weight : 213.0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PY-1551

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?PY-1551
Availability details for
Quantity
  • Availability
  • Add to Cart