COM448635334 Combi-Blocks, Inc.

5-Bromo-N-(2-methoxyethyl)pyridin-2-amine

  • MDL number MFCD11123673
  • Cas Number : 1005010-02-5
  • Molecular Weight : 231.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PY-1559

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?PY-1559
Availability details for
Quantity
  • Availability
  • Add to Cart