COM448635338 Combi-Blocks, Inc.

t-Butyl 2-cyano-2-(5-nitropyridin-2(1H)-ylidene)acetate

  • MDL number MFCD17214297
  • Cas Number : 1255574-90-3
  • Molecular Weight : 263.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Refrigerated
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PY-1564

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?PY-1564
Availability details for
Quantity
  • Availability
  • Add to Cart