COM448635360 Combi-Blocks, Inc.

2-Chloro-6-(4-methylpiperazino)pyridine

  • MDL number MFCD00925758
  • Cas Number : 153976-26-2
  • Molecular Weight : 211.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Refrigerated
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PY-1598

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?PY-1598
Availability details for
Quantity
  • Availability
  • Add to Cart