COM448635365 Combi-Blocks, Inc.

3-Amino-2-hydroxy-6-methylpyridine HCl

  • MDL number MFCD18087751
  • Cas Number : 1257665-17-0
  • Molecular Weight : 160.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PY-1604

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?PY-1604
Availability details for
Quantity
  • Availability
  • Add to Cart