COM448635366 Combi-Blocks, Inc.

3-Amino-5-chloro-2-hydroxypyridine, HCl

  • MDL number MFCD18311909
  • Cas Number : 1261906-29-9
  • Molecular Weight : 181.0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PY-1606

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?PY-1606
Availability details for
Quantity
  • Availability
  • Add to Cart