COM448635373 Combi-Blocks, Inc.

4-Bromo-2-(N-cyclopentylamino)pyridine

  • MDL number MFCD14702599
  • Cas Number : 1209458-99-0
  • Molecular Weight : 241.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PY-1613

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?PY-1613
Availability details for
Quantity
  • Availability
  • Add to Cart