COM448635374 Combi-Blocks, Inc.

N-(4-Bromopyridin-2-yl)formamide

  • MDL number MFCD20491450
  • Cas Number : 1352318-23-0
  • Molecular Weight : 201.0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PY-1617

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?PY-1617
Availability details for
Quantity
  • Availability
  • Add to Cart