COM448635376 Combi-Blocks, Inc.

2-Benzylamino-4-bromopyridine

  • MDL number MFCD14702643
  • Cas Number : 1209457-90-8
  • Molecular Weight : 263.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PY-1620

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?PY-1620
Availability details for
Quantity
  • Availability
  • Add to Cart