COM448635377 Combi-Blocks, Inc.

4-Bromo-N,N-diethylpyridin-2-amine

  • MDL number MFCD11869617
  • Cas Number : 1142194-28-2
  • Molecular Weight : 229.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PY-1621

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?PY-1621
Availability details for
Quantity
  • Availability
  • Add to Cart