COM448635385 Combi-Blocks, Inc.

4-Bromo-2-(4-methylpiperidino)pyridine

  • MDL number MFCD11869628
  • Cas Number : 1142194-45-3
  • Molecular Weight : 255.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Refrigerated
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PY-1640

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?PY-1640
Availability details for
Quantity
  • Availability
  • Add to Cart