COM448635386 Combi-Blocks, Inc.

4-Bromo-N-propylpyridin-2-amine

  • MDL number MFCD18434476
  • Cas Number : 1280786-74-4
  • Molecular Weight : 215.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PY-1648

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?PY-1648
Availability details for
Quantity
  • Availability
  • Add to Cart