COM448635387 Combi-Blocks, Inc.

4-Bromo-2-(N-isopropylamino)pyridine

  • MDL number MFCD14702648
  • Cas Number : 1209458-03-6
  • Molecular Weight : 215.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PY-1649

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?PY-1649
Availability details for
Quantity
  • Availability
  • Add to Cart