COM448635394 Combi-Blocks, Inc.

2,6-Bis(benzyloxy)pyridine

  • MDL number MFCD05863260
  • Cas Number : 16727-46-1
  • Molecular Weight : 291.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PY-1669

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?PY-1669
Availability details for
Quantity
  • Availability
  • Add to Cart