COM448635412 Combi-Blocks, Inc.

2-Propoxyquinoline

  • MDL number MFCD18783197
  • Cas Number : 945-83-5
  • Molecular Weight : 187.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PY-1697

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?PY-1697
Availability details for
Quantity
  • Availability
  • Add to Cart