COM448635424 Combi-Blocks, Inc.

3-Amino-2-bromo-5,6-dichloropyridine

  • MDL number MFCD18783198
  • Cas Number : 1253889-50-7
  • Molecular Weight : 241.9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PY-1714

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?PY-1714
Availability details for
Quantity
  • Availability
  • Add to Cart