COM448635430 Combi-Blocks, Inc.

5-Bromo-1-ethylindazole

  • MDL number MFCD17010084
  • Cas Number : 590417-96-2
  • Molecular Weight : 225.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PY-1720

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?PY-1720
Availability details for
Quantity
  • Availability
  • Add to Cart