COM448635431 Combi-Blocks, Inc.

5-Bromo-2-ethyl-2H-indazole

  • MDL number MFCD18434483
  • Cas Number : 590417-97-3
  • Molecular Weight : 225.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PY-1721

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?PY-1721
Availability details for
Quantity
  • Availability
  • Add to Cart