COM448635438 Combi-Blocks, Inc.

1-(5-Bromo-6-methylpyridin-2-yl)-4-methylpiperazine

  • MDL number MFCD27578176
  • Cas Number : 1454301-28-0
  • Molecular Weight : 270.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Refrigerated
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PY-1728

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?PY-1728
Availability details for
Quantity
  • Availability
  • Add to Cart