COM448635439 Combi-Blocks, Inc.

2-Bromo-5-methoxypyridine 1-oxide

  • MDL number MFCD18914531
  • Cas Number : 1305322-98-8
  • Molecular Weight : 204.0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PY-1730

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?PY-1730
Availability details for
Quantity
  • Availability
  • Add to Cart