COM448635455 Combi-Blocks, Inc.

6-Bromo-2-(N-methylamino)quinazoline

  • MDL number MFCD13191815
  • Cas Number : 882670-77-1
  • Molecular Weight : 238.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PY-1753

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?PY-1753
Availability details for
Quantity
  • Availability
  • Add to Cart