COM448635461 Combi-Blocks, Inc.

6-Chloro-3-methoxypyridazin-4-amine

  • MDL number MFCD00234095
  • Cas Number : 14369-14-3
  • Molecular Weight : 159.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PY-1765

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?PY-1765
Availability details for
Quantity
  • Availability
  • Add to Cart