COM448635477 Combi-Blocks, Inc.

4-Bromo-N-cyclohexylpyridin-2-amine

  • MDL number MFCD18434486
  • Cas Number : 1262293-75-3
  • Molecular Weight : 255.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PY-1810

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?PY-1810
Availability details for
Quantity
  • Availability
  • Add to Cart