COM448635493 Combi-Blocks, Inc.

4,6-Dibromo-5-hydroxypicolinic acid

  • MDL number MFCD18783201
  • Cas Number : 64354-26-3
  • Molecular Weight : 296.9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PY-1835

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?PY-1835
Availability details for
Quantity
  • Availability
  • Add to Cart