COM448635494 Combi-Blocks, Inc.

5-Bromo-2-(4-ethylpiperidino)pyridine

  • MDL number MFCD09910224
  • Cas Number : 364794-57-0
  • Molecular Weight : 270.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PY-1837

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?PY-1837
Availability details for
Quantity
  • Availability
  • Add to Cart