COM448635506 Combi-Blocks, Inc.

2-Phenylpyridine-3-carbonitrile

  • MDL number MFCD02929371
  • Cas Number : 39065-49-1
  • Molecular Weight : 180.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PY-1852

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?PY-1852
Availability details for
Quantity
  • Availability
  • Add to Cart