COM448635508 Combi-Blocks, Inc.

6-Chloro-8-methylquinoline

  • MDL number MFCD10686879
  • Cas Number : 19655-50-6
  • Molecular Weight : 177.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PY-1856

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?PY-1856
Availability details for
Quantity
  • Availability
  • Add to Cart