COM448635510 Combi-Blocks, Inc.

5-Chloro-3-fluoro-2-methylpyridine

  • MDL number MFCD18256290
  • Cas Number : 1210868-68-0
  • Molecular Weight : 145.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PY-1860

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?PY-1860
Availability details for
Quantity
  • Availability
  • Add to Cart