COM448635516 Combi-Blocks, Inc.

2-chloro-6-[2-(pyrrolidin-1-yl)ethoxy]pyridine

  • MDL number MFCD16987542
  • Cas Number : 1242241-03-7
  • Molecular Weight : 226.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PY-1866

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?PY-1866
Availability details for
Quantity
  • Availability
  • Add to Cart