COM448635517 Combi-Blocks, Inc.

6-Nitro-1H-quinolin-2-one

  • MDL number MFCD11501896
  • Cas Number : 64495-55-2
  • Molecular Weight : 190.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PY-1867

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?PY-1867
Availability details for
Quantity
  • Availability
  • Add to Cart