COM448635520 Combi-Blocks, Inc.

Methyl 6-amino-3-(4-t-butylphenyl)picolinate

  • MDL number MFCD18783204
  • Cas Number : 1280786-65-3
  • Molecular Weight : 284.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PY-1870

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?PY-1870
Availability details for
Quantity
  • Availability
  • Add to Cart