COM448635524 Combi-Blocks, Inc.

2-(Cyclopropylmethoxy)-5-fluoropyridine

  • MDL number MFCD18914534
  • Cas Number : 1305322-92-2
  • Molecular Weight : 167.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PY-1874

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?PY-1874
Availability details for
Quantity
  • Availability
  • Add to Cart